Learn More
Invitrogen™ RBL1 Monoclonal Antibody (KAB6)

Description
RBL1 Monoclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | RBL1 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | KAB6 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.08% sodium azide |
| Gene | RBL1 |
| Gene Accession No. | P28749, Q64701 |
| Gene Alias | 107 kDa retinoblastoma-associated protein; AW547426; CP107; DNA-binding protein RFX5; MGC40006; p107; pRb1; RB transcriptional corepressor like 1; Rbl1; Regulatory factor X 5; regulatory factor X, 5 (influences HLA class II expression); regulatory factor X5; retinoblastoma-like 1; retinoblastoma-like 1 (p107); retinoblastoma-like protein 1; RFX5 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.