Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RBM16 Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501559077
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-155-9077 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-155-9077 Supplier Bethyl Laboratories, Inc Supplier No. A301036AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Rat

May play a role in mRNA processing.

Specifications

Antigen RBM16
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene SCAF8
Gene Accession No. Q6DID3, Q9UPN6
Gene Alias Protein SCAF8, CDC5L complex-associated protein 7, RNA-binding motif protein 16, SR-related and CTD-associated factor 8, SCAF8, CCAP7, KIAA1116, RBM16
Gene Symbols Scaf8
Host Species Rabbit
Immunogen Between 1025 and 1075
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 106583, 22828
Target Species Human, Mouse
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.