Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Rbm20 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558068 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558068 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558068 Supplier Invitrogen™ Supplier No. PA558068
Add to Cart
Edge
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ADWGNGRDSW EHSPYARREE ERDPAPWRDN GDDKRDRMDP WAHDRKHHPR QLDKAELDER PEGGRPHREK YPRSGSPNLP HSVSSYKSRE D Highest antigen sequence identity to the following orthologs: Mouse - 70%, Rat - 69%.

This gene encodes a protein that likely binds RNA. Mutations in this gene have been associated with familial dilated cardiomyopathy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Rbm20
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Rbm20
Gene Accession No. Q5T481
Gene Alias 1110018J23Rik; 2010003H22Rik; AI646761; probable RNA-binding protein 20; Rbm20; RNA binding motif protein 20; RNA-binding motif protein 20; RNA-binding protein 20
Gene Symbols Rbm20
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human Rbm20. Recombinant protein control fragment (Product #RP-97212).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 282996
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.