Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RBM24 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566881
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566881 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566881 Supplier Invitrogen™ Supplier No. PA566881
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PALIQRPFGIP AHYVYPQAFV QPGVVIPHVQ PTA Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.

RBM24 is preferentially expressed in muscle during differentiation in vitro and may regulate myogenic differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RBM24
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Rbm24
Gene Accession No. Q9BX46
Gene Alias 6330546B05Rik; AI606861; dJ259A10.1; Rbm24; rbm24a; RNA binding motif protein 24; RNA binding motif protein 24a; RNA binding protein; rna-binding; RNA-binding motif protein 24; RNA-binding protein 24; RNA-binding protein SEB-4; RNA-binding region (RNP1, RRM) containing 6; RNA-binding region-containing protein 6; RNPC6; seb-4
Gene Symbols Rbm24
Host Species Rabbit
Immunogen Recombinant Human RBM24. Recombinant protein control fragment (Product #RP-105410).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 221662
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.