Learn More
RBM25/RED120 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A301067AT
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse
RNA-binding protein that acts as a regulator of alternative pre-mRNA splicing. Involved in apoptotic cell death through the regulation of the apoptotic factor BCL2L1 isoform expression. Modulates the ratio of proapoptotic BCL2L1 isoform S to antiapoptotic BCL2L1 isoform L mRNA expression. When overexpressed, stimulates proapoptotic BCL2L1 isoform S 5′-splice site (5′-ss) selection, whereas its depletion caused the accumulation of antiapoptotic BCL2L1 isoform L. Promotes BCL2L1 isoform S 5′-ss usage through the 5′-CGGGCA-3′ RNA sequence. Its association with LUC7L3 promotes U1 snRNP binding to a weak 5′ ss in a 5′-CGGGCA-3′-dependent manner. Binds to the exonic splicing enhancer 5′-CGGGCA-3′ RNA sequence located within exon 2 of the BCL2L1 pre-mRNA. Also involved in the generation of an abnormal and truncated splice form of SCN5A in heart failure.Specifications
| RBM25/RED120 | |
| Polyclonal | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| P49756 | |
| RBM25 | |
| Between 150 and 200 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot | |
| Unconjugated | |
| RBM25 | |
| arg/Glu/Asp-rich protein of 120 kDa, Arg/Glu/Asp-rich protein, 120 kDa, fSAP94, functional spliceosome-associated protein 94, NET52, Protein S164, RED120, RNA-binding motif protein 25, RNA-binding region (RNP1, RRM) containing 7, RNA-binding region-containing protein 7, RNPC7, S164, Snu71, U1 small nuclear ribonucleoprotein 1SNRP homolog | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 58517 | |
| 4° C | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.