Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RBM26 Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501559800
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-155-9800 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-155-9800 Supplier Bethyl Laboratories, Inc Supplier No. A301216A
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse

6 isoforms of the human protein are produced by alternative splicing. Ota T., Nat. Genet. 36:40-45(2004). Dunham A., Nature 428:522-528(2004). Eichmueller S., Proc. Natl. Acad. Sci. U.S.A. 98:629-634(2001).

Specifications

Antigen RBM26
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.2 mg/ml
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene RBM26
Gene Accession No. Q5T8P6
Gene Alias ARRS2, C13orf10, RP11-255E21.1, SE70-2, ZC3H17, CTCL tumor antigen se70-2, RNA-binding motif protein 26, RNA-binding protein 26, acidic rich RS domain containing 2, cutaneous T-cell lymphoma tumor antigen se70-2
Gene Symbols RBM26
Host Species Rabbit
Immunogen Between 450 and 500
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64062
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.