Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RBM27 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557475
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557475 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557475 Supplier Invitrogen™ Supplier No. PA557475
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LLLQQQQTLS HLSQQHHHLP QHLHQQQVLV AQSAPSTVHG GIQKMMSKPQ TSGAYVLNKV PVKHRLGHAG GNQSDASHLL NQSGGAGEDC QIFS Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 81%.

RBM27 gene ontology annotations related to this gene include mRNA processing; positive regulation of RNA export from nucleus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RBM27
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RBM27
Gene Accession No. Q9P2N5
Gene Alias A730010I06; acidic rich RS domain containing 1; AI043120; ARRS1; Kiaa1311; mKIAA1311; peri implantation stem cell 1; periimplantation stem cell 1; peri-implantation stem cell protein 1; Psc1; RBM27; RNA binding motif protein 27; RNA-binding motif protein 27; RNA-binding protein 27; ZC3H18; ZC3H20
Gene Symbols RBM27
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RBM27. Recombinant protein control fragment (Product #RP-96862).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54439
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.