Learn More
Invitrogen™ RBM3 Polyclonal Antibody

Description
Immunogen sequence: MSSEEGKLFV GGLNFNTDEQ ALEDHFSSFG PISEVVVVKD RETQRSRGFG FITFTNPEHA SVAMRAMNGE SLDGRQIRVD HAGKSARGTR GGGFGAHGRG RSYSRGGGDQ GYGSGRYYDS RPGGYGYGYG RSRDYNGRNQ GGYDRYSGGN YRDNYDN; Positive Samples: HeLa, Jurkat, HL-60; Cellular Location: Cell projection, Cytoplasm, Nucleus, dendrite.
Specifications
Specifications
| Antigen | RBM3 |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.76 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | RBM3 |
| Gene Accession No. | O89086, P98179, Q925G0 |
| Gene Alias | 2600016C11Rik; hypothetical protein LOC509771; IS1-RNPL; OTTHUMP00000025800; OTTHUMP00000025802; putative RNA-binding protein 3; Rbm3; RNA binding motif (RNP1, RRM) protein 3; RNA binding motif protein 3; RNA-binding motif protein 3; RNA-binding protein 3; RNPL |
| Gene Symbols | RBM3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.