Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RBM5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. 501732610 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-2610 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-2610 Supplier Proteintech Group Inc Supplier No. 199301AP150UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

6 isoforms of the human protein are produced by alternative splicing. Ota T., Nat. Genet. 36:40-45(2004). Dunham A., Nature 428:522-528(2004). Eichmueller S., Proc. Natl. Acad. Sci. U.S.A. 98:629-634(2001).

Specifications

Antigen RBM5
Applications Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene RBM5
Gene Accession No. B2GV05, P52756, Q91YE7
Gene Alias G15, H37, LUCA15, Protein G15, RBM5, RMB5, RNA binding motif protein 5, RNA binding protein 5
Gene Symbols Rbm5
Host Species Rabbit
Immunogen RBM5 Fusion Protein Ag13908
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10181, 300996, 83486
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.