Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RBM8A Monoclonal Antibody (4C4), FITC
GREENER_CHOICE

Catalog No. PIMA546552
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA546552 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA546552 Supplier Invitrogen™ Supplier No. MA546552
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Perform heat mediated antigen retrieval with citrate buffer pH 6 before commencing with IHC staining protocol. Western blot detects a band of approximately 21 kDa (predicted molecular weight: 21 kDa).

This gene encodes a protein with a conserved RNA-binding motif. The protein is found predominantly in the nucleus, although it is also present in the cytoplasm. It is preferentially associated with mRNAs produced by splicing, including both nuclear mRNAs and newly exported cytoplasmic mRNAs. It is thought that the protein remains associated with spliced mRNAs as a tag to indicate where introns had been present, thus coupling pre- and post-mRNA splicing events. Previously, it was thought that two genes encode this protein, RBM8A and RBM8B; it is now thought that the RBM8B locus is a pseudogene. Two alternative start codons result in two forms of the protein, and this gene also uses multiple polyadenylation sites.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RBM8A
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 4C4
Concentration 1 mg/mL
Conjugate FITC
Formulation PBS with 0.09% sodium azide
Gene RBM8A
Gene Accession No. Q9DF42, Q9Y5S9
Gene Alias 2310057C03Rik; AA673428; Binder of OVCA1-1; BOV1; BOV-1; BOV1A; BOV-1A; BOV1B; BOV-1B; BOV1C; BOV-1C; C1DELq21.1; DEL1q21.1; fb34f03; HSPC114; MDS014; Rbm8; RBM8A; rbm8-A; rbm8a.S; rbm8a-A; rbm8a-b; RBM8B; ribonucleoprotein RBM8; ribonucleoprotein RBM8A; Ribonucleoprotein RBM8A-A; RNA binding motif protein 8; RNA binding motif protein 8A; RNA binding motif protein 8A S homeolog; RNA binding motif protein 8B; rna-binding; RNA-binding motif protein 8A; RNA-binding motif protein 8A-A; RNA-binding protein 8A; RNA-binding protein 8A-A; RNA-binding protein Y14; TAR; tsunagi; Unknown (protein for MGC:127566); wu:fb34f03; XELAEV_18042903mg; Y14; zgc:110525; ZNRP; zrnp1
Gene Symbols RBM8A, rbm8a.S
Host Species Mouse
Immunogen Human full length native protein (purified).
Purification Method Protein A
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 399167, 9939
Target Species Human, Frog
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.