Learn More
AdipoGen RBP4 ms rat Dual ELISA Kit
SDP
Supplier:  AdipoGen AG45A0012YEKKI01
Encompass_Preferred
Colorimetric Sandwich ELISA Assay. Detects mouse and rat RBP4. Does not cross-react with human RBP4, mouse adiponectin, mouse resistin, mouse Nampt, rat adiponectin, rat resistin or rat Nampt. Colorimetric assay. Sample Types Cell Culture Supernatant, Serum, Urine. Range 0.188 to 12ng/ml. Sensitivity 60pg/ml. Retinol binding protein 4 RBP4 RBP is a 21kDa secreted protein, a member of the lipocalin family and is known as the primary transporter of retinol vitamin A to tissues. A recent report revealed RBP4 as an adipokine linking glucose transporter 4 GLUT4 suppression in adipose tissue to insulin. Elevated human and mouse serum RBP4 levels are associated with insulin resistance and its severity, obesity and certain components of metabolic syndrome. Furthermore, human serum RBP4 levels are closely related to renal function and recent studies have shown an association between serum RBP4 levels and urinary albumin excretion.
Catalog No. 50-256-2743
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.