Learn More
TARGETMOL CHEMICALS INC RBPJ INHIBITOR-1 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T35566
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. RBPJ Inhibitor-1 (RIN1) (RIN1) the RBPJ inhibitor blocks the functional interaction of RBPJ with SHARP. RBPJ Inhibitor-1 (RIN1) inhibits NOTCH-dependent tumor cell proliferation[1]. purity: 99%
Catalog No. 50-258-0213
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.