Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ RBS™ pF Detergent Concentrate, 1 kg PROMO

A phosphate-free, surfactant-based cleaning solution for use on many laboratory surfaces, labware utensils and glassware.

Supplier:  Thermo Scientific™ 27959

Catalog No. PI27959PM


Get 14% Off Protein Preparation Products
View Available Promotions
Only null left
Edge
Add to Cart

Description

Description

Highlights:

  • Ideal for removing radioactivity on contaminated bench surfaces
  • Low-foaming
  • Rinses easily without leaving residues
  • Fully biodegradable
  • Phosphate free
Specifications

Specifications

Liquid
2% (v/v) Solution
1 kg
RBS™
Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 20 to 25°C.
1 kg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only. Not for use in diagnostic procedures.