Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Creative Biolabs Rabbit Anti-T7 RNA Polymerase Antibody, 500 μg, Polyclonal, Liquid, >95% purity
SDP

Supplier:  Creative Biolabs MOB0296MC500UG

Encompass_Preferred

The Creative Biolabs Rabbit Anti-T7 RNA Polymerase Antibody is a high-quality polyclonal antibody developed for the specific detection of recombinant T7 RNA Polymerase. Validated for use in a variety of applications, including immunoblotting and immunoprecipitation, this antibody ensures reliability and precision in molecular biology research.

  • Host is rabbit
  • Clonality is polyclonal
  • Immunogen is full-length T7 RNA Polymerase (recombinant)
  • Isotype is Rabbit IgG
  • Applications include ELISA, Western Blot, dot blot, and immunoprecipitation
  • Purity is >95% as determined by SDS-PAGE
  • Format is liquid
  • Quantity is 500 μg
  • Recommended storage is -20°C or lower (long-term), 2-8°C (short-term)
  • Handling precaution: avoid repeated freeze-thaw cycles
  • Optimal Western Blot dilution is 1:500 to 1:2000

Catalog No. NC2159881


May include imposed supplier surcharges.
Only null left
Add to Cart
This item is not returnable. View return policy

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.