Learn More
Innovative Research Inc Rbt IgG Aff Pur FITC lbl 1mg
Supplier:  Innovative Research Inc IRBIGGAPFITC1MG
Encompass_Preferred
Rabbit IgG Affinity Purified FITC Labeled 1mg - Rabbit IgG Affinity Purified FITC Labeled from Innovative Research has been purified from normal Rabbit Serum using Protein A affinity chromatography then labeled at primary amines with fluorescein isothiocyanate (FITC). This is a frozen liquid buffered in 0.02M Sodium Phosphate; 0.15M NaCl; pH 7.4 and a purity of 95%. This product is useful for ELISA, Western Blot, and as an isotype control. More Details Source Rabbit Serum Purification IgG Fractionated Purity 95% Storage Conditions -70 C
Catalog No. 50-274-4047
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.