Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RC74 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531305
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531305 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531305 Supplier Invitrogen™ Supplier No. PA531305
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: NT2D1, IMR32, U87-MG, MCF-7. Predicted reactivity: Mouse (99%), Zebrafish (84%), Xenopus laevis (87%), Chicken (92%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

INTS9 is a subunit of the Integrator complex, which associates with the C-terminal domain of RNA polymerase II large subunit (POLR2A)and mediates 3-prime end processing of small nuclear RNAs U1 (RNU1)and U2 (RNU2)(Baillat et al., 2005).
TRUSTED_SUSTAINABILITY

Specifications

Antigen RC74
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene INTS9
Gene Accession No. Q9NV88
Gene Alias BC028953; CPSF2L; D14Ertd231e; INT9; integrator complex subunit 9; INTS9; Protein related to CPSF subunits of 74 kDa; RC74; RC-74
Gene Symbols INTS9
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the N-terminus region of human RC74. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55756
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.