Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RCAS1 Mouse anti-Human, Clone: 4H8A12, Proteintech
SDP

Catalog No. 501736604 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-6604 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-6604 Supplier Proteintech Group Inc Supplier No. 661701IG150UL

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

This gene was identified as an estrogen-responsive gene. Regulation of transcription by estrogen is mediated by estrogen receptor which binds to the estrogen-responsive element found in the 5′-flanking region of this gene. The encoded protein is a tumor-associated antigen that is expressed at high frequency in a variety of cancers. Two transcript variants differing in the 5′ UTR, but encoding the same protein, have been identified for this gene.

Specifications

Antigen RCAS1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 4H8A12
Concentration 0.67 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene EBAG9
Gene Accession No. O00559
Gene Alias EB9, EBAG9, PDAF, RCAS1
Gene Symbols EBAG9
Host Species Mouse
Immunogen RCAS1 Fusion Protein Ag2905
Purification Method Protein G
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9166
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.