Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RCBTB1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566040
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566040 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566040 Supplier Thermo Scientific Supplier No. PA566040
Only null left
Add to Cart
Add to Cart

RCBTB1 Polyclonal Antibody

RCBTB1 Polyclonal Antibody for ICC/IF

TRUSTED_SUSTAINABILITY

Specifications

Antigen RCBTB1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RCBTB1
Gene Accession No. Q8NDN9
Gene Alias 5430409I18Rik; AW111883; Chronic lymphocytic leukemia deletion region gene 7 protein; CLL deletion region gene 7 protein; CLLD7; CLLL7; E4.5; GDP/GTP exchange factor (GEF)-like protein; GLP; RCBTB1; RCC1 and BTB domain containing protein 1; RCC1 and BTB domain-containing protein 1; regulator of chromosome condensation (RCC1) and BTB (POZ) domain containing protein 1; regulator of chromosome condensation and BTB domain-containing protein 1; RP11-185C18.1
Gene Symbols RCBTB1
Host Species Rabbit
Immunogen Recombinant Human RCBTB1. Recombinant protein control fragment (Product #RP-105051).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55213
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.