Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RCOR2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110286
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110286 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110286 Supplier Invitrogen™ Supplier No. PA5110286
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HHPLRTRRRP PKGMYLSPEG LTAVSGSPDL ANLTLRGLDS QLISLKRQVQ SMKQTNSSLR QALEGGIDPL RPPEANTKFN SRWTTDEQLL AVQAIRRYGK D.

RCOR2 may act as a component of a corepressor complex that represses transcription.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RCOR2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.87 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene RCOR2
Gene Accession No. Q5FWT8, Q8C796
Gene Alias 1A13; AW122124; co-repressor of Rest; CoREST; M-CoREST; Rcor; Rcor1; Rcor2; RE1-silencing transcription factor (REST) co-repressor 1; RE1-silencing transcription factor (REST) co-repressor 2; REST corepressor 2; REST co-repressor 2
Gene Symbols RCOR2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 250-350 of human RCOR2 (NP_775858.2).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 104383, 305811
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.