Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RD Plastics Clear Reclosable Zip Bags
SDP

Catalog No. 01818100
Change view
Click to view available options
Dimensions (L x W):
15 x 22.5 cm (6 x 9 in.)
20.32 x 30.32 cm (8 x 10 in.)
30 x 37.5 cm (12 x 15 in.)
30.4 x 30.4 cm (12 x 12 in.)
60.9 x 60.9 cm (24 x 24 in.)
Thickness (English):
2 mil
4 mil
Light gauge
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W) Thickness (English)
01-818-100 15 x 22.5 cm (6 x 9 in.) 4 mil
01-818-101 30 x 37.5 cm (12 x 15 in.) 4 mil
19-130-6043 20.32 x 30.32 cm (8 x 10 in.) 2 mil
19-165-620 30.4 x 30.4 cm (12 x 12 in.) 4 mil
01-817-100 30 x 37.5 cm (12 x 15 in.) 2 mil
19-130-4163 60.9 x 60.9 cm (24 x 24 in.) Light gauge
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 01-818-100 Supplier RD Plastics Supplier No. C21
Add to Cart
Add to Cart

For non-biohazard use

  • Constructed of heavy weight 4 mil polyethylene
  • Feature reclosable zip seal

Specifications

Color Clear
For Use With (Application) Non-biohazard use, general purpose
Material Polyethylene
Thickness (English) 4 mil
Dimensions (L x W) 15 x 22.5 cm (6 x 9 in.)
Product Type General Purpose Bag
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.