Learn More
RD Plastics Reclosable Specimen Transport Bag

Description
- Specimen transport
- Two wall or three wall
- Biohazard print
- Score line for easy opening
- Absobent pad
Specifications
Specifications
| Closure Type | Reclosable |
| Color | Clear |
| For Use With (Application) | For specimen transport |
| Includes | 3-wall Construction, Storage Instructions |
| Thickness (English) | 2 mil |
| Dimensions (L x W) | 9 x 6 in. |
| Length (English) | 9 in. |
| Width (English) | 6 in. |
| Product Type | Reclosable Specimen Transport Bag |
| Length (Metric) | 22.8 cm |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.