Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RD Plastics Color Coded Ziplock Biohazard Specimen Bags
SDP

Catalog No. 23700232
Change view
Click to view available options
Color:
Blue
Green
Red
Yellow
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
23-700-232 Green
23-700-231 Yellow
23-700-234 Red
23-700-233 Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 23-700-232 Supplier RD Plastics Supplier No. Q865
Add to Cart
Add to Cart

Opaque colored specimen transport bags feature a reclosable seal with a tear away top for easy opening. Available in yellow, green, blue or red.

  • LDPE bags in 3 Wall style with 2 mil thickness
  • Printed with black "SPECIMEN" and biohazard markings
  • Red option also printed with "STAT"
  • Color coded for easy specimen handling protocols
  • Includes a paperwork pouch

Specifications

Autoclavable Not Autoclavable
Closure Type Tear-away Zip-top
Color Green
For Use With (Application) Specimen transport
Includes Biohazard print, paperwork pouch
Material LDPE
No. per Case 1000
No. per Pack 100
Thickness (English) 2 mil
Dimensions (L x W) 9 x 6 in. (22.8 x 15.2cm)
Length (English) 9 in.
Width (English) 6 in.
Product Type Transport Bag
Capacity (English) 1.5 lb.
Length (Metric) 22.8 cm
Width (Metric) 15.2 cm
Thickness (Metric) 0.279 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.