Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RDH10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543997
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543997 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543997 Supplier Invitrogen™ Supplier No. PA543997
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: QSNEETAGMV RHIYRDLEAA DAAALQAGNG EEEILPHCNL QVFTYTCDVG Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.

RDH10 generates all-trans retinal from all-trans retinol and may plan an important role in the photic visual cycle. All-trans retinal is isomerized to 11-cis retinal by the retinal G protein-coupled receptor (RGR; MIM 600342) when the retinal pigment epithelium (RPE) is illuminated.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RDH10
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene RDH10
Gene Accession No. Q8IZV5
Gene Alias 3110069K09Rik; 4921506A21Rik; AI875664; AW549993; D1Ertd762e; m366Asp; RDH10; Retinol dehydrogenase 10; retinol dehydrogenase 10 (all-trans); SDR16C4; Short chain dehydrogenase/reductase family 16C member 4; short chain dehydrogenase/reductase family 16C, member 4; UNQ9375/PRO34191
Gene Symbols RDH10
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human RDH10 (aa 71-120).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 157506
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.