Learn More
Andwin Scientific Re202 Rotary Evaporator Set With Bm302 Water Bath And Diagonal Condenser A 115V
Supplier: Andwin Scientific RE202AWA
Standard rotary evaporator with slide jack mechanism for convenient height adjustment. Set includes BM302-A water bath (115V) and glassware set A with diagonal condenser. Market leader in rotation speed range at 5~315 rpm. Designed with set inversion function where glassware and bath can be set on either side, left or right, depending on user’s dominant hand and installation location. Standard equipped with condenser insulation kit to suppress condensation on the condenser’s surface. Available in 220V and in combination with oil bath.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.