Learn More
Thermo Scientific™ Reacti-Therm™ Magnetic Stirrers for Reacti-Vial™ Small Reaction Vials
Catalog No. p-6775055
Change view
Click to view available options
For Use With (Equipment):
0.3 and 1mL Reacti-Vials
3, 5, and 10 mL Reacti-Vials
Description:
6 Pack
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Description
PI16010 0.3 and 1mL Reacti-Vials 6 Pack
PI16000 3, 5, and 10 mL Reacti-Vials 6 Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PI16010 Supplier Thermo Scientific™ Supplier No. TS16010
Only null left
Add to Cart
Add to Cart
Use these triangular stir bars for faster reaction times and smooth mixing in vials of the Thermo Scientific™ Reacti-Therm™ Derivatization System.
Use these triangular stir bars for faster reaction times and smooth mixing of small samples in conical-bottom vials of the Thermo Scientific™ Reacti-Therm™ Derivatization System. The stir bars provide simultaneous stirring of samples in multiple vials when used with a Reacti-Therm Heating/Stirring Module equipped with appropriate Reacti-Block Aluminum Blocks.
TRUSTED_SUSTAINABILITY
Specifications
Description 6 Pack
For Use With (Equipment) 0.3 and 1mL Reacti-Vials
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.