Learn More
Aat Bioquest ReadiUse probenecid 25 mM stabilized aqueous solution (10x10 mL)
SDP
Supplier:  Aat Bioquest 20062
Encompass_Preferred
Probenecid is an inhibitor of organic-anion transporters located in cell membranes. This catalog is for 10x10 mL.
Catalog No. 50-224-9805
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.