Learn More
Sartorius Octet Residual Protein A Detection Kit, Accurate Quantitation of Leached Protein A, Compatible with Octet Systems, Kit
Catalog No. NC1147499 Shop All Sartorius Products
Encompass
The Octet Residual Protein A (RPA) Detection Kit provides a rapid and efficient method for quantitating leached Protein A in bioprocess samples. Unlike traditional ELISA methods, it features a streamlined dip-and-read workflow without the need for boiling or centrifugation, allowing for quick analysis in less than 2 hours. Designed for use in research and manufacturing environments, it accommodates 96 samples at once and is compatible with most Octet systems.
Catalog No. NC1147499 Supplier Sartorius Supplier No. 185128
The Octet Residual Protein A (RPA) Detection Kit provides a rapid and efficient method for quantitating leached Protein A in bioprocess samples. Unlike traditional ELISA methods, it features a streamlined dip-and-read workflow without the need for boiling or centrifugation, allowing for quick analysis in less than 2 hours. Designed for use in research and manufacturing environments, it accommodates 96 samples at once and is compatible with most Octet systems.