Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ready-Made LB (Luria-Bertani) Broth Powder

Catalog No. aaj7585230
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ7585230 250 g
AAJ75852A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ7585230 Supplier Thermo Scientific Supplier No. J7585230
Only null left
Add to Cart

For propagation of bacteria

For the propagation of bacteria.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Off-white free flowing powder
  • pH(2.0%, H2O): 7.0 ± 0.2

Contents

  • Casein Peptone: 50%
  • Yeast Extract: 25%
  • Sodium Chloride: 25%
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Form Powder
Product Type Dehydrated Culture Media
For Use With (Application) Propagation of Bacteria
Packaging Type Plastic Bottle
Quantity 250 g

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.