Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Ready-Made Terrific Broth Powder

Catalog No. AAJ75856A1 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ75856A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ75856A1 Supplier Thermo Scientific Chemicals Supplier No. J75856A1
Add to Cart

missing translation for 'validMainframeMsg'

Provides greater yields of plasmid DNA and recombinant proteins than LB media by allowing for greater cell density

  • Off-white free flowing powder
  • pH(5.08%, H2O): 7.2 ± 0.2

Contents

  • Casein Peptone: 12g/L
  • Yeast Extract: 24g/L
  • Potassium Phosphate Dibasic: 12.5g/L
  • Potassium Phosphate Monobasic: 2.3g/L
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Form Powder
Product Type Microbiology Media
For Use With (Application) Provides Greater Yields of Plasmid DNA and Recombinant Proteins
Packaging Type Plastic Bottle
Quantity 1 kg

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.