Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Ready-To-Go™ GenomiPhi HY DNA Amplification Kit

Catalog No. 45003104 Shop All Cytiva Products
Change view
Click to view available options
No. of Reactions:
24
96
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
45-003-104 24
45003105 96
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 45-003-104 Supplier Cytiva Supplier No. 25660324
Add to Cart
Add to Cart

illustra Ready-To-Go GenomiPhi HY DNA Amplification Kit offers highly efficient and representative whole-genome amplification with 40 to 60++g yield from nanogram amounts of DNA sample.

illustra Ready-To-Go GenomiPhi HY DNA Amplification Kit is part of the Phi29 DNA polymerase family of products from Cytiva.

  • Representative amplification of the whole genome.
  • Preformulated, predispensed, single-dose, lyophilized cakes for rapid reaction set-up and greater reproducibility.
  • Quick and simple automation-friendly protocol; no thermal cycler required.
  • Shipping and storage at room temperature provides easy access and reduced carbon footprint.
  • Yields high-quality DNA (e.g. for high-throughput genotyping and DNA archival).

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store the kit at ambient temperature.Once opened, completely reseal the pouch containingthe Ready-To-Go GenomiPhi V3 DNA amplification kit cakes. To ensure maximum shelf life and optimal performance, storage in a desiccator is recommended.Store recons
Description illustra Ready-To-Go GenomiPhi HY DNA Amplification Kit offers highly efficient and representative whole-genome amplification with 40 to 60 μg yield from nanogram amounts of DNA sample.
For Use With (Application) Genomic DNA Amplification
Includes 1 x 2.6mL 2X Denaturation Buffer, 3 x 8-well strips Ready-To-Go™ GenomiPhi HY cakes, 1 x vial control DNA, 12 strips of 8 domed caps
No. of Reactions 24
Product Type illustra Ready-To-Go GenomiPhi HY DNA Amplification Kit 24 Reactions
Size > 10 kb
Test Time 4 h
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.