Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Ready-to-Go™ You-Prime First-Strand Beads

Catalog No. 45001416 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
50 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-001-416 50 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45-001-416 Supplier Cytiva Supplier No. 27926401
NLSU: Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Fifty preformulated, single-dose first-strand reaction mix beads prepackaged in white, thin-walled 0.5mL tubes compatible with most thermal cyclers. Each tube contains two beads containing buffer, dATP, dCTP, dGTP, dTTP, murine reverse transcriptase, RNase inhibitor, and RNase/DNase-free BSA. Also includes five control mix beads packaged in red tubes containing globin mRNA, buffer, and two globin-specific PCR primers for evaluating the performance of the Reaction Mix Beads and the PCR amplification.

Use for synthesis of first-strand cDNA templates from total RNA or polyadenylated RNA using a primer of choice. Cytiva Life Sciences™ Ready-to-Go™ You-Prime First-Strand Beads are perfect for RT-PCR applications.

  • Predispensed, ambient-temperature-stable, single-dose reaction beads minimize pipetting errors, avoid cross-contamination, and ensure optimal performance
  • Each first-strand reaction requires only the addition of RNA in DEPC-treated water and a user-selected first-strand primer
  • First-strand reaction may be primed with oligo(dT) primers, random primers such as pd(N)6, or custom primers complementary to a specific mRNA sequence
  • Conditions of this reaction have been optimized to permit full-length transcription of RNAs 7kb or more in length
  • Final reaction volume is 33μL once reconstituted
  • Ideal for research applications that use PCR to detect and quantify eukaryotic RNA from a variety of samples

TRUSTED_SUSTAINABILITY

Specifications

Description The kit provides 50 preformulated, single-dose reaction beads prepackaged in thin-walled 0.5 mL tubes compatible with most thermal cyclers. For synthesis of first-strand cDNA templates from total RNA or polyadenylated RNA using a primer of choice.
For Use With (Application) First-Strand
Includes Fifty preformulated, single-dose first-strand reaction mix beads prepackaged in white, thin-walled 0.5mL tubes compatible with most thermal cyclers, each tube contains two beads containing Buffer, dATP, dCTP, dGTP, dTTP, murine reverse transcriptase, RNase inhibitor and RNase/DNase-free BSA
No. of Reactions 50
Product Type Ready-To-Go You-Prime First-Strand Beads
Quantity 50 Reactions
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.