Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Reagent Alcohol 200 Proof, Greenfield

Catalog No. 502946015
Encompass
Change view
Click to view available options
Packaging:
4x1 Gallon Case
Metal Drum
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging
50-294-6015 Poly Pail
50-294-6033 Metal Drum
50-294-6004 4x1 Gallon Case
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 50-294-6015 Supplier Thermo Scientific Production Chemicals and Services Supplier No. P241000200PL05
Add to Cart
Add to Cart

Specifications

Quantity 5 Gallon
Packaging Poly Pail
Boiling Point 172.4 °F (78 °C)
Percent Purity 200 Proof
Grade ACS
Chemical Name or Material Reagent Alcohol 200 Proof
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.