Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Reagent Alcohol, ACS Reagent, for organic synthesis, prep-LC, and general laboratory use, anhydrous, SDA 3A, 90 ±1%, Solstice

Catalog No. 6000560 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
19 L
4 x 4 L
Packaging:
Metal Can
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-005-60 19 L Metal Can
60-005-62 4 x 4 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-005-60 Supplier Solstice Supplier No. LP09019
Add to Cart
Add to Cart

Chemical Identifiers

CAS 64-17-5 , 67-56-1 , 67-63-0
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.069
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Synonym ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
PubChem CID 702
ChEBI CHEBI:16236
IUPAC Name ethanol
SMILES CCO

Specifications

CAS Max % 90.0000%, 5.0000%, 5.0000%
Density 0.7847 g/cm3
Quantity 19 L
Flash Point 15°C
Packaging Metal Can
Assay Percent Range 94 to 96%
Water 0.5% max.
UN Number UN1987
Beilstein 1718733
Boiling Point 78.32°C
Solubility Information Miscible in Water at 20°C
Grade ACS
Chemical Name or Material Reagent Alcohol, ACS Reagent, for organic synthesis, prep-LC, and general laboratory use, anhydrous, SDA 3A, 90 ±1%
Show More Show Less
ShelfLife 547 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Alcohols, n.o.s.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.