Learn More
Specifications
Specifications
| Color | Colorless |
| CAS Max % | 66.39, 30.25, 1.68, 1.68 |
| Decomposition Information | Carbon monoxide; Carbon dioxide; May release flammable gases |
| Density | 0.95g/mL |
| Quantity | 4 L |
| Identification | Passes Test |
| Packaging | Poly Bottle |
| Physical Form | Liquid |
| Specific Gravity | 0.95g/mL +/- 0.008 |
| Linear Formula | C2H6O |
| Show More |
Safety and Handling
| Hazard Category | Danger |
| Hazard Statement | GHS H Statement Flammable liquid and vapor. Causes skin irritation. Causes serious eye irritation. May cause drowsiness or dizziness. Suspected of damaging fertility or the unborn child. Causes damage to organs (central nervo |
| Precautionary Statement | GHS P Statement Obtain special instructions before use. Do not handle until all safety precautions have been read and understood. Keep away from heat, hot surfaces, open flames, sparks. No smoking. Keep container tightly closed. |
| EINECSNumber | 200-578-6 |
| Recommended Storage | Room Temperature |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.