Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Reagent Alcohol, CHROMASOLV™, for HPLC, Solstice

Catalog No. 60045752 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-045-752 1 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-045-752 Supplier Solstice Supplier No. 2707411L

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 64-17-5 , 67-56-1 , 67-63-0
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.07
MDL Number MFCD00003568
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Synonym ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
PubChem CID 702
ChEBI CHEBI:16236
IUPAC Name ethanol
SMILES CCO

Specifications

Absorbance max. 0.35 at 220 nm, max. 0.65 at 210 nm, max. 1.00 at 205 nm, max. 0.01 at 400 nm, max. 0.01 at 300 nm, max. 0.04 at 255 nm
CAS Max % 90.0000%, 5.0000%, 5.0000%
Density 0.7847 g/cm3
Quantity 1 L
Flash Point 15°C
Packaging Glass Bottle
UN Number UN1987
Beilstein 1718733
Boiling Point 78.32°C
Percent Purity >89%
Grade HPLC
Chemical Name or Material Reagent Alcohol, CHROMASOLV™, for HPLC
Show More Show Less
ShelfLife 547 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Alcohols, n.o.s.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.