Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Reagent Alcohol, CHROMASOLV™, for HPLC, Solstice
GSA_VA

Catalog No. 6000559 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
4 x 4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-005-59 4 x 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-005-59 Supplier Solstice Supplier No. 2707414X4L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

  • For HPLC and organic synthesis
  • High-purity, multipurpose solvents
  • High lot-to-lot consistency

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-17-5 , 67-56-1 , 67-63-0
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.07
MDL Number MFCD00003568
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Synonym Ethyl alcohol, Reagent alcohol, RGA, Denatured alcohol
IUPAC Name ethanol
SMILES CCO

Specifications

CAS Max % 90.00, 5.00, 5.00
Density 0.785 g/cm3
Quantity 4 x 4 L
Flash Point 15°C.The physical data is that of the main component.
Packaging Glass Bottle
Physical Form Liquid
Boiling Point 78°C
Grade HPLC
Hazard Category Danger
Hazard Statement H370+H319+H225
Precautionary Statement P210+P233+P240+P241+P242+P243+P260+P264+P270+P280
Recommended Storage Room Temperature
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.