Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Reagent Alcohol (Denatured Alcohol), 20% (v/v), Ricca Chemical
SDP

Catalog No. 01213111 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
4 L
10 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-213-111 4 L
01-213-116 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-213-111 Supplier Ricca Chemical Company Supplier No. R25462004A
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7732-18-5 , 64-17-5 , 67-63-0 , 67-56-1 , 67-56-1
Molecular Formula H2O
Molecular Weight (g/mol) Mixture
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
IUPAC Name water
SMILES O

Specifications

Melting Point Approximately 0°C
Color Colorless
CAS Min % 83.33%, 0.75%, 15.08%, 0.83%
Appearance Colorless Liquid
CAS Max % 0.75%, 83.33%, 0.83%, 15.08%
Quantity 4 L
Packaging 4 L HDPE Bottle
Physical Form Liquid
Specific Gravity 20% (v/v)
Boiling Point Approximately 100°C
Percent Purity 20%
Grade Laboratory
Chemical Name or Material Reagent Alcohol, 20% (V/V)
Show More Show Less
ShelfLife 24 Months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.