Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Reagent Alcohol (Denatured Alcohol), 20% (v/v), Ricca Chemical
SDP

Catalog No. p-200004037
Click to view available options
Quantity:
1 L
4 L
10 L
55 gal.

Chemical Identifiers

CAS 7732-18-5, 64-17-5, 67-63-0, 67-56-1, 67-56-1
Molecular Formula H2O
Molecular Weight (g/mol) Mixture
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
IUPAC Name water
SMILES O

Specifications

Melting Point Approximately 0°C
Color Colorless
CAS Min % 83.33%, 0.75%, 15.08%, 0.83%
Appearance Colorless Liquid
CAS Max % 0.75%, 83.33%, 0.83%, 15.08%
Boiling Point Approximately 100°C
Quantity 1 L
Packaging 1 L HDPE Bottle
Physical Form Liquid
Specific Gravity 20% (v/v)
Percent Purity 20%
Grade Laboratory
Chemical Name or Material Reagent Alcohol, 20% (V/V)
Show More Show Less
ShelfLife 24 Months
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.