Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Reagent Alcohol (Denatured Alcohol), 70% (v/v), Ricca Chemical
SDP

Catalog No. 25467025 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
1 L
3.8 L
4 L
10 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
Safety-coated Glass Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
25467025 10 L Cubitainer
25467032 1 L Natural Poly Bottle
25-467-01SC 4 L Safety-coated Glass Bottle
2546705 20 L Cubitainer
R254670038 3.8 L Natural Poly Bottle
25-467-01 4 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 25467025 Supplier Ricca Chemical Company Supplier No. 2546.702.5
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Reagent Alcohol (Denatured Alcohol), 70% (v/v)

  • Reagent Alcohol (Denatured Alcohol), 70% (v/v)

Chemical Identifiers

CAS 64-17-5 , 7732-18-5 , 67-63-0 , 67-56-1
Molecular Weight (g/mol) Mixture
PubChem CID 702
ChEBI CHEBI:16236

Specifications

Melting Point -50°C
Color Colorless
CAS Min % 62.0, 29.4, 3.43, 3.09
CAS Max % 65.0, 30.6, 3.57, 3.21
Quantity 10 L
Flash Point 13°C (55.47°F)
Packaging Cubitainer
Physical Form Liquid
Molecular Formula C2H6O
Synonym ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
Boiling Point 93°C
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
SMILES CCO
IUPAC Name ethanol
Percent Purity 70%
Grade Laboratory
Concentration or Composition (by Analyte or Components) 70% (v/v)
Chemical Name or Material Alcohol, Reagent, Denatured Alcohol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.