Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Reagent Alcohol, Denatured, Exceeds A.C.S. Specifications, HPLC Grade, 94.0-96.0%, Spectrum™ Chemical
SDP

Catalog No. 18613032 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
4 L
200 L
Packaging:
Amber Glass Bottle
Drum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-613-032 200 L Drum
18-613-031 4 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-613-032 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. HP612200LTBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 64-17-5 , 67-56-1 , 67-63-0

Specifications

CAS Max % 80 to 100%, 5 to 10%, 5 to 10%
Quantity 200 L
Packaging Drum
Residue after Evaporation 4 ppm
Water 0.001
Boiling Point 78°C
Percent Purity 94 to 96%
Grade ACS/HPLC
Chemical Name or Material Reagent Alcohol

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.