Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Reagent Alcohol, Denatured, Reagent, ACS, 94-96%, Spectrum™ Chemical
SDP

Catalog No. 18606046 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
6 x 500 mL
4 x 4 L
6 x 1 L
200 L
Packaging:
Amber Glass Bottle
Drum
Poly Bottle
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-606-046 500 mL Amber Glass Bottle
18-606-047 500 mL Poly Bottle
18-606-045 1 L Amber Glass Bottle
18-606-048 6 x 500 mL Amber Glass Bottle
18-606-049 6 x 500 mL Poly Bottle
18-606-050 4 L Amber Glass Bottle
18-606-054 4 L Poly Bottle
18-606-051 6 x 1 L Amber Glass Bottle
18-606-052 4 x 4 L Amber Glass Bottle
18-606-053 4 x 4 L Poly Bottle
18-606-055 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Catalog No. 18-606-046 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. A1040500MLGL
Add to Cart
Add to Cart

A1040, 64-17-5, C2H5OH

Spectrum™ Chemical Reagent Alcohol, Denatured, Reagent, ACS is also known as ethanol and is an extremely volatile and a flammable liquid that is colorless. In the laboratory, it is used mainly as a analytical solvent. As an ACS grade reagent, this Spectrum Chemical manufactured compound is used as the quality standard against which other substances are graded and has met the toughest regulatory standards for quality and pureness.

Chemical Identifiers

CAS 64-17-5 , 67-63-0

Specifications

CAS Max % 0.95, 0.05
Quantity 500 mL
Packaging Amber Glass Bottle
Residue after Evaporation 1e-005
Water 0.005
Boiling Point 78°C
Percent Purity 94 to 96%
Grade Reagent
Chemical Name or Material Reagent Alcohol
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.