Learn More
Med Vet International HbA1c Test Kit DCA™ Systems HbA1c Test, CLIA Waived, 10311134, 10 Tests/Kit, 4 Kits
Supplier:  Med Vet International 213834CS
Encompass_Preferred
Only 1 µL of whole blood needed for HbA1c testing Results in 6 minutes No sample or reagent preparation needed allowing for simple and efficient operation
Catalog No. 50-304-4991
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.