Learn More
Description
Refills for sulfide tests, potassium iodide, chlorine, ferroine indicators and more
Specifications
Specifications
| Volume (Metric) | 500 mL |
| Includes | Mixed acid nitrogen reducing reagent, alkalinity reagent B, chlorine reagent 1 |
| Storage Requirements | 25°C |
| Type | Ferroin Indicator |
| Volume (English) | 16.9 oz. |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

