Learn More
Thomas Scientific Reagent Reservoir, 100 ml, Sterile, Individually Wrapped
SDP
Supplier:  Thomas Scientific 1228K24
Encompass_Preferred
Choose from four styles - 25 mL Divided, 25 mL , 50 mL and 100 mL “V” bottom design allows for maximum recovery of reagents Graduated with convenient pouring spouts at all four corners Extra wide base for increased stability Unique “Nesting” feature makes separation of bulk packed reservoirs easy 25ml divided reservoirs are divided into 3 separate 5ml channels; great for low volume pipetting. Sterile and certified version are certified RNase and DNase-free and non-pyrogenic. Bulk packed reservoirs come in packs of 5 reservoirs.
Catalog No. NC0528909