Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Simport™ Scientific Reagent Reservoir Cover

Catalog No. 01670666 Shop All Simport Scientific Products
Change view
Click to view available options
No. per Pack:
10 Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. per Pack
01-670-666 10 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 01-670-666 Supplier Simport™ Scientific Supplier No. R22515
Add to Cart
Add to Cart
This item is not returnable. View return policy

For labs involved with fraction collecting, compound storage, plate replication, and mixing applications. Suitable for a range of liquid handling configurations from 8 or 12-channel pipettes to 96 or 384-pipette heads. Ideal for small-volume robotic tips used in high-throughput instruments.

  • Robot friendly and designed for simple, economical handling of reagents in automated systems
  • Conforms to ANSI/SLAS standard dimensions
  • Compatible with virtually any automated liquid handler, stacker, and manual procedure
  • Bottom designed to enable funneling of liquid, maximizing reagent recovery with pipettes
  • Low-profile, chemical and heat resistant
  • Can be stored at temperatures as low as -80°C
  • Can be gamma irradiated
  • Certified DNase-, RNase- and pyrogen-free
  • Made from natural virgin polypropylene

Specifications

Dimensions (L x W x H) 127.8 x 85 x 9 mm
Material Polystyrene
Sterility Non-sterile
No. per Case 300 Cs.
No. per Pack 10 Pk.
Autoclavable Not Autoclavable
Disposable Single-use
Bottom Shape Flat
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.