Learn More
Hach Company Chloride Reagent Set, Drop Count Titration
Supplier:  Hach Company 2437600
Encompass
For determination of chloride by argentometric drop-count titration. Replacement reagent set for 8-P Test Kit (144001). Set includes: Chloride 2 Indicator Powder Pillows and Silver Nitrate Titrant (0.0493 N titrant). Approx. 100 tests.
Catalog No. 50-199-5992