Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ Reagents For Bacterial Transformation: Amp, X-GAL, pGAL
SDPSCI_ED

Catalog No. S05680 Shop All EDVOTEK Products
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S05680 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S05680 Supplier Edvotek™ Supplier No. 617

missing translation for 'validMainframeMsg'

For rapid preparation of bacterial cultures for transformation.

  • Includes: AMP, X-Gal, and pGal™ (Does not include media and plasticware)

Specifications

Content And Storage Freezer
For Use With (Application) Rapid preparation of bacterial cultures for transformation
Includes AMP, X-Gal and pGal
Quantity 1 Ea.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.