Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ Reagents For Bacterial Transformation: Bacterial Plating Agar
SDPSCI_ED

Catalog No. s11201b Shop All EDVOTEK Products
Change view
Click to view available options
:
Bacterial Plating Agar
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S11201B Bacterial Plating Agar
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S11201B Supplier Edvotek™ Supplier No. 612

For rapid preparation of bacterial cultures for transformation.

  • 1.05 oz. (30g)

Specifications

Content And Storage Room Temperature
Product Type Agar
For Use With (Application) Rapid preparation of bacterial cultures for transformation
Quantity 30 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.