Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ Reagents For Bacterial Transformation: Readypour Luria Broth
SDPSCI_ED

Catalog No. S11201E Shop All EDVOTEK Products
Change view
Click to view available options
Includes:
With Ampicillin
Without Ampicillin
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
S11201E Without Ampicillin
S11201F With Ampicillin
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S11201E Supplier Edvotek™ Supplier No. 615
Add to Cart
Add to Cart

For rapid preparation of bacterial cultures for transformation.

  • 170 mL

Specifications

Content And Storage Room Temperature
Form Agar Suspension
Manufacturing Origin United States
Product Type Tubed Media
Sterility Sterile
Target Organism Class Bacteria
Test Type Microbial Identification
For Use With (Application) Rapid preparation of bacterial cultures for transformation
Includes Without Ampicillin
Quantity 170 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.