Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ Reagents For Bacterial Transformation: X-gal
SDPSCI_ED

Catalog No. S11201D Shop All EDVOTEK Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S11201D 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S11201D Supplier Edvotek™ Supplier No. 614
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For rapid preparation of bacterial cultures for transformation.

  • 8.8 oz. (250mg)

Specifications

Content And Storage Store below 0°C.
Format Tube
Promoter None (Promoterless)
For Use With (Application) Rapid preparation of bacterial cultures for transformation
Product Type X-gal
Quantity 250 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.